missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%

Brand:  Alfa Aesar™ H63889.03

Code : NEW

Additional Details : CAS Number : 1212136-00-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498566

  • £130.60 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Description

Description

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
Specifications

Chemical Identifiers

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
237.27
tert-Butyl (R)-4-Methyl-2,2-dioxo-1,2,3-oxathiazolidine-3-carboxylate
1212136-00-9
MFCD09864994

Specifications

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
Solid
237.27
100° to 107°C
tert-Butyl (R)-4-Methyl-2,2-dioxo-1,2,3-oxathiazolidine-3-carboxylate
1212136-00-9
97%
MFCD09864994
Off-white to pale brown
1g
Videos
SDS