missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog Number 11966481
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1212734-83-2
C6H10F3N
153.15
NAXDEFXCCITWEU-RXMQYKEDSA-N
r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine
42580053
(2R)-2-(trifluoromethyl)piperidine
C1CCNC(C1)C(F)(F)F
This item is not returnable. View return policy
1212734-83-2
C6H10F3N
153.15
NAXDEFXCCITWEU-RXMQYKEDSA-N
r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine
42580053
(2R)-2-(trifluoromethyl)piperidine
C1CCNC(C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 80.0°C (200.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material (R)-2-(Trifluoromethyl)piperidine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-2-(Trifluoromethyl)piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'