missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylglycinol, 98%, Thermo Scientific Chemicals

Catalog Number p-3737204
44.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56613-80-0
C8H12NO
138.19
MFCD00008062
IJXJGQCXFSSHNL-MRVPVSSYSA-O
r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol
2724025
(2R)-2-amino-2-phenylethanol
[NH3+][C@H](CO)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56613-80-0
C8H12NO
138.19
MFCD00008062
IJXJGQCXFSSHNL-MRVPVSSYSA-O
r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol
2724025
(2R)-2-amino-2-phenylethanol
[NH3+][C@H](CO)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 79°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(NH2)CH2OH
Quantity 5 g
Fieser 17,279
Specific Rotation Condition −30.50° (20°C c=1,1N HCl)
Specific Rotation -30.5
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2-Phenylglycinol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-2-Phenylglycinol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'