missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3211047
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
938-79-4
C10H12O2
164.20
MFCD00063165
OFJWFSNDPCAWDK-UHFFFAOYNA-N
r---2-phenylbutyric acid, 2r-2-phenylbutanoic acid, 2-phenylbutyric acid, r, r-2-phenylbutyric acid, benzeneacetic acid, alpha-ethyl-, r, r-2-phenylbutanoic acid, 2r-2-phenyl-butanoic acid, 2r-2-phenyl-mutanoic acid, r---2-phenylbutanoic acid, benzeneacetic acid,a-ethyl-, ar
785330
(2R)-2-phenylbutanoic acid
CCC(C(O)=O)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
938-79-4
C10H12O2
164.20
MFCD00063165
OFJWFSNDPCAWDK-UHFFFAOYNA-N
r---2-phenylbutyric acid, 2r-2-phenylbutanoic acid, 2-phenylbutyric acid, r, r-2-phenylbutyric acid, benzeneacetic acid, alpha-ethyl-, r, r-2-phenylbutanoic acid, 2r-2-phenyl-butanoic acid, 2r-2-phenyl-mutanoic acid, r---2-phenylbutanoic acid, benzeneacetic acid,a-ethyl-, ar
785330
(2R)-2-phenylbutanoic acid
CCC(C(O)=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0550g/mL
Boiling Point 130°C (3.0 mbar)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C2H5CH(C6H5)CO2H
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Quantity 1 g
Beilstein 09,IV,1815
Specific Gravity 1.055
Solubility Information Solubility in water: insoluble
Formula Weight 164.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Phenylbutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-2-Phenylbutyric acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'