missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Methyl-CBS-oxazaborolidine monohydrate, 94%, Thermo Scientific™
£24.34 - £64.17
Chemical Identifiers
CAS | 112022-83-0 |
---|---|
Molecular Formula | C18H20BNO |
Molecular Weight (g/mol) | 277.174 |
MDL Number | MFCD00078440 |
InChI Key | VMKAFJQFKBASMU-QGZVFWFLSA-N |
Synonym | r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst |
PubChem CID | 9838490 |
IUPAC Name | (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
SMILES | B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387296
|
Thermo Scientific Alfa Aesar
L09230.MD |
250mg |
£24.34
250mg |
Estimated Shipment: 07-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11377296
|
Thermo Scientific Alfa Aesar
L09230.03 |
1g |
£64.17
1g |
Estimated Shipment: 10-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
112022-83-0 | |
277.174 | |
VMKAFJQFKBASMU-QGZVFWFLSA-N | |
9838490 | |
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C |
C18H20BNO | |
MFCD00078440 | |
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst | |
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
Specifications
(R)-2-Methyl-CBS-oxazaborolidine monohydrate | |
C18H20BNO | |
9059874 | |
250mg | |
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C | |
277.174 | |
295.19 (277.17 Anhydrous) | |
94% |
112022-83-0 | |
MFCD00078440 | |
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst | |
VMKAFJQFKBASMU-QGZVFWFLSA-N | |
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole | |
9838490 | |
+74° (c=1 in Methanol) |