missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Methyl-CBS-oxazaborolidine monohydrate, 94%, Thermo Scientific™

Catalog Number 11377296
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
112022-83-0
C18H20BNO
277.17
MFCD00078440
VMKAFJQFKBASMU-QGZVFWFLSA-N
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
9838490
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
[H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
112022-83-0
C18H20BNO
277.17
MFCD00078440
VMKAFJQFKBASMU-QGZVFWFLSA-N
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
9838490
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
[H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 9059874
Optical Rotation +74° (c=1 in Methanol)
Formula Weight 295.19 (277.17 Anhydrous)
Percent Purity 94%
Chemical Name or Material (R)-2-Methyl-CBS-oxazaborolidine monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'