missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Fmoc-amino)butyric acid, 98%, Thermo Scientific Chemicals

Catalog Number 15420899
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
170642-27-0
C19H19NO4
325.364
MFCD00270336
XQIRYUNKLVPVRR-QGZVFWFLSA-N
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
853017
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
170642-27-0
C19H19NO4
325.364
MFCD00270336
XQIRYUNKLVPVRR-QGZVFWFLSA-N
fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
853017
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 325.36
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (R)-2-(Fmoc-amino)butyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'