missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Benzyloxypropionic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7058473
28.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100836-85-9
C10H12O3
180.203
MFCD06799065
XWAVPOFYNPXXEL-MRVPVSSYSA-N
r-2-benzyloxy propanoic acid, r-+-2-benzyloxy propionic acid, r-+-2-benzyloxypropionic acid, o-benzyl-d-lactic acid, 2r-2-benzyloxy propanoic acid, propanoic acid, 2-phenylmethoxy-, 2r, r-benzyloxypropionic acid, r-2-benzyloxy-propionic acid, 2r-2-benzyloxy propionic acid, 2r-2-phenylmethoxypropanoic acid
7000104
(2R)-2-phenylmethoxypropanoic acid
CC(C(=O)O)OCC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100836-85-9
C10H12O3
180.203
MFCD06799065
XWAVPOFYNPXXEL-MRVPVSSYSA-N
r-2-benzyloxy propanoic acid, r-+-2-benzyloxy propionic acid, r-+-2-benzyloxypropionic acid, o-benzyl-d-lactic acid, 2r-2-benzyloxy propanoic acid, propanoic acid, 2-phenylmethoxy-, 2r, r-benzyloxypropionic acid, r-2-benzyloxy-propionic acid, 2r-2-benzyloxy propionic acid, 2r-2-phenylmethoxypropanoic acid
7000104
(2R)-2-phenylmethoxypropanoic acid
CC(C(=O)O)OCC1=CC=CC=C1

(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 51°C to 53°C
Quantity 5 g
Beilstein 4675524
Solubility Information Insoluble in water.
Optical Rotation +90° (c=1 in IMS)
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material (R)-(+)-2-Benzyloxypropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'