missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-2-Benzyloxypropionic acid, 98%, Thermo Scientific Chemicals
28.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Melting Point | 51°C to 53°C |
Quantity | 5 g |
Beilstein | 4675524 |
Solubility Information | Insoluble in water. |
Optical Rotation | +90° (c=1 in IMS) |
Formula Weight | 180.2 |
Percent Purity | 98% |
Chemical Name or Material | (R)-(+)-2-Benzyloxypropionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'