missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 97%

Brand:  Alfa Aesar™ H62328.03

Code : NEW

Additional Details : CAS Number : 214852-61-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15455555

  • £88.93 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

(R)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
553.7
Z-D-Dab(Boc)-OH.DCHA; Nα-Benzyloxycarbonyl-Nγ-Boc-D-2,4-diaminobutyric acid dicyclohexylammonium salt
214852-61-6
MFCD01318750

Specifications

(R)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
Solid
553.7
1g
214852-61-6
97%
MFCD01318750
Z-D-Dab(Boc)-OH.DCHA; Nα-Benzyloxycarbonyl-Nγ-Boc-D-2,4-diaminobutyric acid dicyclohexylammonium salt
Videos
SDS