missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 363160010

£108.84 valid until 30/06/2017
Log in to get the promotional price



UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 374790-95-1 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10436062

  • £272.10 / 1g


EU Stock 1
Add to basket

Specifications Description & Specifications

Chemical Identifiers

(R)-2-Aminobut-3-en-1-ol, benzoate salt
≥94%
C11H15NO3
MFCD01631086
95%, (98% e.e.)
95%
209.24

Specifications

(R)-2-Aminobut-3-en-1-ol, benzoate salt
97.5% min. (GC)
Glass bottle
≥94%
White to almost white
209.24
MFCD01631086
95%, (98% e.e.)
Authentic
Crystalline Powder
95%
C11H15NO3
1g
Documentation

Technical Documents

Special Offers

Special Offers