missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 363165000

Code : 29

Additional Details : CAS Number : 374790-95-1 Weight : 0.05050kg

Packaging Glass bottle
Quantity 500mg

 View more versions of this product

Product Code. 10120952

  • £63.90 / 500mg


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

(R)-2-Aminobut-3-en-1-ol, benzoate salt
95%
C11H15NO3
MFCD01631086
95%, (98% e.e.)
95%
209.24

Specifications

(R)-2-Aminobut-3-en-1-ol, benzoate salt
374790-95-1
Authentic
Crystalline Powder
95%
White to almost white
209.24
MFCD01631086
95%, (98% e.e.)
97.5% min. (GC)
Glass bottle
95%
90.5°C
C11H15NO3
500mg
Videos
SDS