missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-butanol, 98%, Thermo Scientific Chemicals

Catalog Number p-3777216
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
5856-63-3
C4H11NO
89.14
MFCD00064419
JCBPETKZIGVZRE-UHFFFAOYNA-N
r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r
2723856
2-aminobutan-1-ol
CCC(N)CO
This item is not returnable. View return policy
5856-63-3
C4H11NO
89.14
MFCD00064419
JCBPETKZIGVZRE-UHFFFAOYNA-N
r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r
2723856
2-aminobutan-1-ol
CCC(N)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -2°C
Color White
Density 0.9470g/mL
Boiling Point 172°C to 174°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CH2OH
Refractive Index 1.4495 to 1.4535
Quantity 25 mL
Beilstein 04,III,771
Merck Index 10, 429
Specific Gravity 0.947
Specific Rotation Condition −10° (19°C c=neat)
Specific Rotation -10
Solubility Information Solubility in water: miscible. Other solubilities: very soluble in most of the usual solvents,hardly soluble in aliphatic hydrocarbons
Formula Weight 89.14
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material (R)-(-)-2-Amino-1-butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'