missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H62283.06

Code : NEW

Additional Details : CAS Number : 201472-66-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477246

  • £161.40 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

(R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
485.66
Boc-Dab(Boc)-OH.DCHA; (R)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
201472-66-4
MFCD00238312

Specifications

(R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
Solid
485.66
5g
201472-66-4
95%
MFCD00238312
Boc-Dab(Boc)-OH.DCHA; (R)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
Videos
SDS