missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Diamino-1,1'-binaphthalene, 99%, ACROS Organics™

Brand:  Acros Organics 356512500

Code : 29

Additional Details : CAS Number : 18741-85-0 Weight : 0.05025kg


Packaging Glass bottle

 View more versions of this product

Product Code. 10454345

  • £61.10 / 250mg


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

(R)-(+)-2, 2'-Diamino-1, 1'-binaphthalene
98.5
99%
C20H16N2
MFCD00145204
18741-85-0
100.0
99%
284.36

Specifications

(R)-(+)-2, 2'-Diamino-1, 1'-binaphthalene
Off-white to brown
250mg
98.5
Authentic
Solubility in water: slightly soluble in water.
Powder
99%
284.36
156.00 (20.00°C c=1,pyridine)
242.0° to 244.0°C
18741-85-0
100.0
Glass bottle
156
99%
C20H16N2
MFCD00145204
Videos
SDS