missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 95+%, ACROS Organics™

Catalog Number p-3523240
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
99646-28-3
C48H40P2
678.796
MFCD01311709
IOPQYDKQISFMJI-UHFFFAOYSA-N
r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
4189906
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C
This item is not returnable. View return policy
99646-28-3
C48H40P2
678.796
MFCD01311709
IOPQYDKQISFMJI-UHFFFAOYSA-N
r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
4189906
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C
CAS Min % 95.0
CAS Max % 100.0
Melting Point 255.0°C to 257.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition +167.50° (20°C c=0,3%,toluene)
Specific Rotation + 167.50
Solubility Information Solubility in water: insoluble.
Formula Weight 678.79
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material (R)-(+)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'