missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethylamine, 99+%, produced by BASF AG, Thermo Scientific Chemicals

Catalog Number p-2689245
40.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
3886-69-9
MFCD00064405
RQEUFEKYXDPUSK-SSDOTTSWSA-N
r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
643189
CHEBI:35322
(1R)-1-phenylethanamine
CC(C1=CC=CC=C1)N
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
3886-69-9
MFCD00064405
RQEUFEKYXDPUSK-SSDOTTSWSA-N
r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
643189
CHEBI:35322
(1R)-1-phenylethanamine
CC(C1=CC=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless
Density 0.9500g/mL
Boiling Point 184.0°C to 186.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N
Linear Formula C6H5CH(CH3)NH2
Refractive Index 1.5250 to 1.5270
Quantity 50 g
Beilstein 12,II,586
Fieser 09,363; 03,199; 14,257; 13,185; 12,319; 11,411; 07,393; 05,441; 02,271; 01,838; 06,457
Merck Index 13,6054
Specific Gravity 0.95
Specific Rotation Condition + 40.00 (20.00°C neat)
Specific Rotation + 40.00
Solubility Information Solubility in water: 40g/L water (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Molecular Weight (g/mol) 121.18
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Phenylethylamine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(+)-1-Phenylethylamine, produced by BASF AG
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'