missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-BOC-3-(Aminomethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4228391
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
140645-23-4
C11H22N2O2
214.31
WPWXYQIMXTUMJB-SECBINFHSA-N
r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502023
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN
This item is not returnable. View return policy
140645-23-4
C11H22N2O2
214.31
WPWXYQIMXTUMJB-SECBINFHSA-N
r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502023
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 214.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-1-BOC-3-(Aminomethyl)piperidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'