missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.0200g/mL |
Boiling Point | 108°C to 115°C (5.0 mmHg) |
Flash Point | 140°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Quantity | 1 g |
Specific Gravity | 1.02 |
Show More |
Safety and Handling
Product Identifier
- (R)-(-)-1-Benzyl-3-aminopyrrolidine
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'