missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ACROS Organics™

Brand:  Acros Organics 437890050

Code : 29

Additional Details : CAS Number : 114715-39-8 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 2b Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 10492580

  • £215.70 / 5g


Estimated Shipment
23-03-2018
Add to basket


Specifications

Chemical Identifiers

(R)-(-)-1-Benzyl-3-aminopyrrolidine
98.5
≥98.5%
C11H16N2
114715-39-8
100.0
99%
176.26

Specifications

(R)-(-)-1-Benzyl-3-aminopyrrolidine
98.5
Liquid
99%
176.26
-1° to -3° (20°C, 589nm) (neat)
Clear colorless to yellow
114715-39-8
100.0
≥98.5%
C11H16N2
Authentic
1.02g/mL
5g
Videos
SDS