missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine, 97%, Thermo Scientific Chemicals

Catalog Number 15478476
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
300543-56-0
C17H19ClN2
286.803
MFCD11519277
UZKBSZSTDQSMDR-QGZVFWFLSA-N
r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
668697
1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
This item is not returnable. View return policy
300543-56-0
C17H19ClN2
286.803
MFCD11519277
UZKBSZSTDQSMDR-QGZVFWFLSA-N
r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
668697
1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

Solubility
Soluble in chloroform, methanol, and dichloromethane. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Soluble in chloroform,methanol,and dichloromethane. Slightly soluble in water.
Formula Weight 286.8
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'