missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(R)-(+)-1-(4-Methoxyphenyl)ethylamine use in diastereo-and enantioselective Michael addition reactions. Thiazoles has been prepared from (R)-(+)-1-(4-Methoxyphenyl)ethylamine.
Solubility
Soluble in water (10g/L).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
(R)-(+)-1-(4-Methoxyphenyl)ethylamine use in diastereo-and enantioselective Michael addition reactions. Thiazoles has been prepared from (R)-(+)-1-(4-Methoxyphenyl)ethylamine.
Solubility
Soluble in water (10g/L).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
Specifications
Melting Point | <-20°C |
Density | 1.02 |
pH | 11 |
Boiling Point | 65°C (0.38 mmHg) |
Odor | Amine-like |
Refractive Index | 1.533 |
Quantity | 5 g |
UN Number | UN2735 |
Beilstein | 2413029 |
Sensitivity | Air Sensitive |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'