missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,1'-Binaphthyl-2,2'-diylhydrogenphosphate, 90%, Maybridge

Brand:  Maybridge MO07668DA

Code : 31

Additional Details : CAS Number : 39648-67-4 Weight : 0.05100kg Transport : UN number : 3464 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10402633

  • £10.00 / 1g


Estimated Shipment date
from Supplier 19-01-2018
Add to basket

Specifications

Chemical Identifiers

(R)-(-)-1, 1'-Binaphthyl-2, 2'-diylhydrogenphosphate
90%
348.29
90%
C20H13O4P

Specifications

(R)-(-)-1, 1'-Binaphthyl-2, 2'-diylhydrogenphosphate
Amber Glass Bottle
90%
348.29
1g
39648-67-4
90%
C20H13O4P
- 607.50 (20.00°C c=1.4,CH3OH)
Videos
SDS