missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate), 97%, Thermo Scientific™

Catalog Number p-66803
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
126613-06-7
C22H12F6O6S2
550.44
MFCD00274615
OYJLCOSEYYZULE-UHFFFAOYSA-N
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
389883
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
126613-06-7
C22H12F6O6S2
550.44
MFCD00274615
OYJLCOSEYYZULE-UHFFFAOYSA-N
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
389883
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 80°C to 85°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CF3SO3C10H6-)2
Quantity 250 mg
Specific Rotation Condition - 149.00 (20.00°C c=1,CHCl3)
Specific Rotation -149
Solubility Information Solubility in water: insoluble
Formula Weight 550.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(-)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'