missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals

Catalog Number 10782841
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18531-94-7
C20H14O2
286.32
MFCD00004068
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
11762
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
This item is not returnable. View return policy
18531-94-7
C20H14O2
286.32
MFCD00004068
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
11762
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 205°C to 211°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC10H6C10H6OH
Quantity 250 mg
Merck Index 15,123
Specific Rotation + 35.5
Solubility Information Solubility in water: insoluble
Formula Weight 286.32
Percent Purity 99%
Chemical Name or Material (R)-(+)-1, 1'-Bi-2-naphthol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-1,1'-Bi-2-naphthol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'