missing translation for 'onlineSavingsMsg'
Learn More

(-)-Quinpirole hydrochloride, Tocris Bioscience™

Selective D2-like agonist

Brand:  Tocris (Bio-Techne) 106150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458413

  • £440.72 / 50mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Selective dopamine D2 receptor agonist (Ki values are 4.8, ∽24, ∽30 and 1900 nM at D2, D3, D4 and D1 receptors respectively).
Specifications

Chemical Identifiers

85798-08-9
255.79
--quinpirole hydrochloride, quinpirole hydrochloride, quinpirole hcl, unii-t6i2w5v2k1, quinpirole hydrochloride usan, dsstox_cid_25519, dsstox_rid_80926, dsstox_gsid_45519, --4ar,8ar-4,4a,5,6,7,8,8a,9-octahydro-5-propyl-1h-pyrazolo 3,4-g quinoline monohydrochloride, quinpirole hydrochloride usan
(4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride
C13H22ClN3
HJHVRVJTYPKTHX-HTMVYDOJSA-N
55397
CCCN1CCCC2C1CC3=C(C2)NN=C3.Cl

Specifications

(-)-Quinpirole hydrochloride
85798-08-9
--quinpirole hydrochloride, quinpirole hydrochloride, quinpirole hcl, unii-t6i2w5v2k1, quinpirole hydrochloride usan, dsstox_cid_25519, dsstox_rid_80926, dsstox_gsid_45519, --4ar,8ar-4,4a,5,6,7,8,8a,9-octahydro-5-propyl-1h-pyrazolo 3,4-g quinoline monohydrochloride, quinpirole hydrochloride usan
CCCN1CCCC2C1CC3=C(C2)NN=C3.Cl
255.79
255.79
50mg
C13H22ClN3
HJHVRVJTYPKTHX-HTMVYDOJSA-N
(4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride
55397
>98%
Videos