Learn More
(-)-Quinpirole hydrochloride, Tocris Bioscience™
Selective D2-like agonist
POA
Description
Description
Selective dopamine D2 receptor agonist (Ki values are 4.8, ∽24, ∽30 and 1900 nM at D2, D3, D4 and D1 receptors respectively).