missing translation for 'onlineSavingsMsg'
Learn More

Quinolinium dichromate, 97%, Thermo Scientific™

Catalog Number 15401518
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
56549-24-7
C18H16Cr2N2O7
476.325
MFCD00059108
PYEIMZCGCWSHSV-UHFFFAOYSA-P
quinolinium dichromate, acmc-209lt3
11465689
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
This item is not returnable. View return policy
56549-24-7
C18H16Cr2N2O7
476.325
MFCD00059108
PYEIMZCGCWSHSV-UHFFFAOYSA-P
quinolinium dichromate, acmc-209lt3
11465689
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 164°C to 167°C (decomposition)
Color Orange to Yellow
Quantity 25 g
UN Number UN3087
Formula Weight 476.33
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Quinolinium dichromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'