missing translation for 'onlineSavingsMsg'
Learn More

Quinolin-2-amine, 97%, Maybridge

Brand:  Maybridge BTB10673DA

Code : 31

Additional Details : CAS Number : 580-22-3 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10723373

  • £37.08 / 1g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Quinolin-2-amine
96.5
97%
C9H8N2
MFCD00038079
580-22-3
100.0
97%
144.18
2-Quinolinamine

Specifications

Quinolin-2-amine
96.5
Amber Glass Bottle
97%
144.18
2-Quinolinamine
580-22-3
100.0
97%
C9H8N2
MFCD00038079
1g
Videos
SDS