Learn More
Thermo Scientific Maybridge Quinolin-2-amine, 5g, 97%
Brand: Thermo Scientific Maybridge BTB10673DE
Quinolin-2-amine, 5g, 97%
Code : 31
Additional Details : CAS Number : 580-22-3 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge BTB10673DE
Quinolin-2-amine, 5g, 97%
Code : 31
Additional Details : CAS Number : 580-22-3 Weight : 0.05500kg