missing translation for 'onlineSavingsMsg'
Learn More

Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine, Thermo Scientific Chemicals

Catalog Number p-7080778
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
130-95-0
C20H24N2O2
324.42
MFCD00198096
LOUPRKONTZGTKE-ZCXAXGJJNA-N
1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol
129316724
[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
This item is not returnable. View return policy
130-95-0
C20H24N2O2
324.42
MFCD00198096
LOUPRKONTZGTKE-ZCXAXGJJNA-N
1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol
129316724
[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼177°C (decomposition)
Quantity 10 g
Beilstein 91867
Sensitivity Light Sensitive
Merck Index 14,8061
Solubility Information Soluble in alcohol,chloroform and diethyl ether. Slightly soluble in water and glycerol.
Optical Rotation −166° (c=1 in ethanol)
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Formula Weight 324.42
Percent Purity 99%
Assay (total base)
Chemical Name or Material Quinine, Anhydrous, may cont. up to 5% dihydroquinine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'