missing translation for 'onlineSavingsMsg'
Learn More

Quinidine, 98%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3779545
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
56-54-2
C20H24N2O2
324.42
MFCD00135581,MFCD00198096
LOUPRKONTZGTKE-LHHVKLHASA-N
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
441074
CHEBI:28593
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
This item is not returnable. View return policy
56-54-2
C20H24N2O2
324.42
MFCD00135581,MFCD00198096
LOUPRKONTZGTKE-LHHVKLHASA-N
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
441074
CHEBI:28593
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 170.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 8176
Specific Rotation + 256.00
Solubility Information Solubility in water: 1g/2000mL (20°C)- 1g/800mL (100°C). Other solubilities: 1g/36mL alcohol- soluble in methanol,1g/56mL ether- 1g/1.6mL chloroform,practically insoluble in petroleum ether
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
Formula Weight 324.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Quinidine
Show More Show Less
compliance-icons
Product Identifier
  • Quinidine, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'