missing translation for 'onlineSavingsMsg'
Learn More

Quinaldoyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number 11996501
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
50342-01-3
C10H6ClNO
191.61
MFCD02683554
WFVMVMAUXYOQSW-UHFFFAOYSA-N
quinaldoyl chloride, 2-quinolinecarbonyl chloride, quinaldyl chloride 97, quinaldyl chloride, chloroformylquinoline, 2-quinolinecarbonyl chloride 9ci, quinoline-2-carboxylic acid chloride
2760547
quinoline-2-carbonyl chloride
ClC(=O)C1=NC2=CC=CC=C2C=C1
This item is not returnable. View return policy
50342-01-3
C10H6ClNO
191.61
MFCD02683554
WFVMVMAUXYOQSW-UHFFFAOYSA-N
quinaldoyl chloride, 2-quinolinecarbonyl chloride, quinaldyl chloride 97, quinaldyl chloride, chloroformylquinoline, 2-quinolinecarbonyl chloride 9ci, quinoline-2-carboxylic acid chloride
2760547
quinoline-2-carbonyl chloride
ClC(=O)C1=NC2=CC=CC=C2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94°C to 97°C
Color Orange to Brown
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 94% min. (GC)
Formula Weight 191.62
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Quinaldoyl chloride
compliance-icons
Product Identifier
  • Quinaldoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'