Learn More
Quickfit™ PTFE Screwthread Washers
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
Brand: Quickfit QW24/9
Code : 01
Additional Details : Weight : 0.02000kg
Description
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
- For use with Quickfit™ glass screwthreads
- Melamine screwcaps with aperture at the top
- Silicone rubber rings with PTFE washers
- To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap.
Specifications
Washer | |
PTFE | |
For Quickfit™ Glass screw threads | |
24 |