Learn More
Quickfit™ Unbadged Joints Cones
Free from any ceramic marking
£53.00 - £300.00
Product Code | Brand | Cone Size | Thickness (Metric) Wall | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Thickness (Metric) Wall | Price | Quantity & Availability | ||||
12939363
|
Quickfit
CNB10UB |
10/19 | 1mm |
£61.45
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
12468816
|
Quickfit
CNB14UB |
14/23 | 1.5mm |
£53.00
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10830251
|
Quickfit
CNB19UB |
19/26 | 1.8mm |
£56.55
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12614911
|
Quickfit
CNB29UB |
29/32 | 2mm |
£70.65
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
12488816
|
Quickfit
CNB24UB |
24/29 | 1.8mm |
£60.05
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
12458826
|
Quickfit
CNB7UB |
7/16 | 1.5mm |
£81.30
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10735671
|
Quickfit
CNB40UB |
40/38 | 2mm |
£53.60
Pack of 5 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12653012
|
Quickfit
CNB45UB |
45/40 | 2.30mm |
£67.35
Pack of 5 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
13431829
|
Quickfit
CNB34UB |
34/35 | 2.00mm |
£88.30
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15597760
|
Quickfit
CNB12UB |
12/21 | 1.5mm |
£300.00
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15507770
|
Quickfit
CNB55UB |
55/44 | 5mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Free from any ceramic marking
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Manufactured to accuracy which surpasses requirements of ISO 383
- Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
- Cones manufactured with shank length of 100mm