Quickfit™ Screwthread Adapter to Fit Cone
 Secure thermometers and tubes to glass containers.  The  QuickFitTM screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.  
Overview
Brand: Quickfit
Manufacturer Part Number: ST54/28
UNSPSC: 41121800
Code: Z4
Additional Details:
Additional Details: Weight: 0.05000kg
Product Code. 11369083
Call for latest availability
+44 (0)1509 555500
Add to basket
Description and Specification
Specification
Product Type Cone screw thread adapter
Material Pyrex Borosilicate Glass
For Use With (Application) Ideal for securing thermometers, gas inlet tubes
Thread Size 28
Includes Plastic cap, Silicone Rubber ring and PTFE washer
For Use With (Equipment) Screw thread joint
  • With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
  • Ideal for securing thermometers, gas inlet tubes, etc