Learn More
Quickfit™ Plain Short Stem Adapter
Feed distillate from condensers into receiving vessels. QuickFit™> plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
Brand: Quickfit RA1/13
Code : 01
Additional Details : Weight : 0.05000kg
Description
- With short or long stem
- Useful for feeding distillate from condensers into receiving vessels
Specifications
Feeding distillate from condensers into receiving vessels | |
24/29 | |
Receiver adapter |
200mm | |
Borosilicate Glass |