missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Folded, Grade 3 m/N

Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Folded, Grade 3 m/N are suitable for analytical and technical purposes where medium fast filtering is desirable.

Brand:  Sartorius Stedim Biotech™ FT-4-305-110

Additional Details : Weight : 0.06200kg

 View more versions of this product

Product Code. 15952990

  • £37.84 / Pack of 100

Estimated Shipment: 11-02-2022
Add to basket

Description

Description

  • Made of refined pulp and linters
  • Medium fast filtering
  • Non-sterile and bulk packed
Specifications

Specifications

Folded
Cotton Linters and Refined Pulp (>95% alpha-cellulose)
0.14 mm
65 g/m2
7 to 10μm
110 mm
3 m/N
100/Pk.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers