missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Discs, Grade 3 m/N
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Discs, Grade 3 m/N are suitable for analytical and technical purposes where medium fast filtering is desirable.
Brand: Sartorius Stedim Biotech™ FT-3-305-320
Additional Details : Weight : 0.52300kg
Description
- Made of refined pulp and linters
- Medium fast filtering
- Non-sterile and bulk packed
Specifications
65 g/m2 | |
Cotton Linters, Refined Pulp | |
30 sec. | |
100/Pk. | |
320 mm |
Disc | |
7 to 10 μm | |
3 m/N | |
0.14 mm |