Learn More
Sartorius Qualitative Grade 1291 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-209-185
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10674522
  • £36.30 / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
180 sec.
100/Pk.
185 mm
2 to 3 μm
1291
0.16 mm