Learn More
Alfa Aesar™ QuadraPure™ IDA, 350-750 micron, metal scavenger
Metal Scavenger: Fe, Al, Ga, In, Cu, V, Pb, Ni, Zn, Cd, Be, Mn, Co, Sr, Ba
Brand:  Alfa Aesar™ 045952.06
Code : NEW
Additional Details : Weight : 0.00500kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Quantity 5g
 View more versions of this product
Product Code. 15442587
  • £48.30 / 5g
Estimated Shipment date
from Supplier 24-05-2018
Add to basket
Specifications
Chemical Identifiers
QuadraPure™ IDA metal scavenger
0.73
UN3175
135620-93-8
MFCD07785598
QuadraPure™ Iminodiacetate
Specifications
QuadraPure™ IDA metal scavenger
Beads
MFCD07785598
QuadraPure™ Iminodiacetate
White to pale cream
135620-93-8
0.73
UN3175
350 to 750 Micron
5g