missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ QuadraPure™ C, 0.3-0.8mm, metal scavenger

Brand:  Alfa Aesar™ 046297.A1

Code : NEW

Additional Details : Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15482637

  • £242.80 / 1kg


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Description

Description

  • High-capacity, carbon-based adsorbent in the form of spherical particles used for scavenging of trace organic impurities
  • Free-flowing and free of dust, QuadraPure™ C is easily removed from reaction mixtures by filtration
Specifications

Chemical Identifiers

QuadraPure™ C
7440-44-0
0.3 to 0.8mm, metal scavenger
MFCD09753896

Specifications

QuadraPure™ C
7440-44-0
MFCD09753896
1kg
0.3 to 0.8mm, metal scavenger
Beads
Black
Videos
SDS