Learn More
Alfa Aesar™ QuadraPure™ AMPA, 350-750 micron, metal scavenger
Metal Scavenger: Fe, Cu, Ni, V, Al, Co
Brand:  Alfa Aesar™ 045917.06
Code : NEW
Additional Details : Weight : 0.00500kg Transport : UN number : 3077 Chem class : 1 Pack group : III
Quantity 5g
 View more versions of this product
Product Code. 15412567
  • £43.04 / 5g
Estimated Shipment date
from Supplier 20-02-2018
Add to basket
Specifications
Chemical Identifiers
QuadraPure™ AMPA metal scavenger
MFCD07785597
UN3077
QuadraPure™ Aminomethylphosphonic acid
Specifications
QuadraPure™ AMPA metal scavenger
MFCD07785597
QuadraPure™ Aminomethylphosphonic acid
White to beige
Beads
UN3077
350 to 750 Micron
5g