Learn More
Alfa Aesar™ QuadraPure™ AMPA, 350-750 micron, metal scavenger
Metal Scavenger: Fe, Cu, Ni, V, Al, Co
Brand: Alfa Aesar™ 045917.14
Code : NEW
Additional Details : Weight : 0.02500kg Transport : UN number : 3077 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
QuadraPure™ AMPA metal scavenger | |
MFCD07785597 | |
UN3077 | |
QuadraPure™ Aminomethylphosphonic acid |
Specifications
QuadraPure™ AMPA metal scavenger | |
MFCD07785597 | |
QuadraPure™ Aminomethylphosphonic acid | |
White to beige |
Beads | |
UN3077 | |
350 to 750 Micron | |
25g |