missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Julabo™ Quad Distributing Adapter
Quad distributing adapter with barbed fittings
Brand: Julabo™ 8970524
Code : A0
Additional Details : Weight : 0.50000kg
Description
- Quad distributing adapter with barbed fittings
Specifications
25.4mm Tubing | |
Quad Distributing Adapter | |
31.75 mm |