missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Suprasil™ Quartz Ultra-Micro Cell Cuvettes

Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Ultra-Micro Cell Cuvettes come with either a PE stopper or open with pipette tips.

Brand:  Hellma™ 105-202-15-40

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10456719

  • £323.00 / Each


Estimated Shipment
21-05-2018
Add to basket

Description

Description

  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
Specifications

Specifications

50μL
45mm
12.5mm
Ultra Macro Absorption Cells
PE stopper
10mm
QS Quartz Suprasil
Videos
Documentation