missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ QR Racks for Grant Heated Circulating Baths
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments QR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
£100.70
Product Code | Brand | Capacity (English) | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Holds | Price | Quantity & Availability | ||||
11496467
|
Grant Instruments™
QR-24 |
10 tubes | 24 mm Tubes |
£100.70
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11486467
|
Grant Instruments™
QR-19 |
16 Tubes | 16 to 19 mm Tubes |
£100.70
Each |
In Stock
Log in to see stock. |
||||
11476467
|
Grant Instruments™
QR-13 |
30 Tubes | 10 to 13 mm Tubes |
£100.70
Each |
In Stock
Log in to see stock. |
||||
11416477
|
Grant Instruments™
QR-SE |
35 Tubes | 1.5 mL Tubes |
£100.70
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11426477
|
Grant Instruments™
QR-LE |
44 Tubes | 0.5 mL Tubes |
£100.70
Each |
In Stock
Log in to see stock. |
||||
11406477
|
Grant Instruments™
QR-30 |
5 Tubes | 30 mm Tubes |
£100.70
Each |
In Stock
Log in to see stock. |
||||
Description
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Grant Heated Circulating Baths |