missing translation for 'onlineSavingsMsg'
Learn More

Qa-2, FITC, clone: 69H1-9-9, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5996-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268329

  • £49.41 / 50µg


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 69H1-9-9 monoclonal antibody reacts with the mouse Qa-2 molecule, a GPI-linked MHC class Ibeta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level is different depending on the laboratory mouse strain.Applications Reported: The 69H1-9-9 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 69H1-9-9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Qa-2
69H1-9-9
FITC
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Qa2
IgG2a
RUO
Primary
Murine
Videos