missing translation for 'onlineSavingsMsg'
Learn More

Qsonica Q700/Q500 Accessories: 8 Tube Holder Rack

An excellent solution for securely holding tubes in place when working with Q500/Q700 Sonicators.

Brand:  Qsonica 451

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15390372

  • £245.00 / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

  • Holds 8 tubes of 1.5mL
  • Available for either Evergreen or Eppendorf tubes
Specifications

Specifications

Tube Holder Rack
Green
Cup Horn (Part # 431C2)
8 x 1.5 mL Polypropylene Eppendorf Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers