missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvic aldehyde dimethyl acetal, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
Solubility
Soluble in water. 350 g/L at 20°C.
Notes
Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
Solubility
Soluble in water. 350 g/L at 20°C.
Notes
Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -57°C |
Density | 0.998 |
Boiling Point | 133°C to 135°C |
Flash Point | 34°C (93°F) |
Refractive Index | 1.398 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 1560557 |
Solubility Information | Soluble in water. 350g/L at 20°C. |
Formula Weight | 118.13 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'